siRNA Details
Gene Symbol: HIST1H3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003530.4 | NP_003521.2 | 2 | 523 | ||
GenBank mRNA | AK316611.1 | - | 2 | 523 | ||
BC031333.1 | - | 2 | 442 | 3608479 | ||
BQ051491.1 | - | 2 | 466 | |||
DB242999.1 | - | 2 | 523 |
