siRNA Details
Gene Symbol: KLF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016270.3 | NP_057354.1 | 3 | 1037 | ||
GenBank mRNA | AF134053.1 | - | 3 | 939 | ||
AF205849.1 | - | 3 | 1023 | |||
BC071983.1 | - | 3 | 1026 | 5480246 | ||
BQ581981.1 | - | 2 | 73 |
