siRNA Details
Gene Symbol: PRELID3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142405.1 | NP_001135877.1 | 5, 6 | 556 | ||
NM_001142406.1 | NP_001135878.1 | 4, 5 | 580 | |||
NM_006553.3 | NP_006544.2 | 5, 6 | 556 | |||
GenBank mRNA | AK056046.1 | - | 5, 6 | 523 | ||
AK303325.1 | - | 4, 5 | 580 | |||
AL359942.1 | - | 3, 4 | 538 | |||
BC106750.2 | - | 5, 6 | 513 | 40022464 | ||
CR627465.1 | - | 3, 4 | 538 | |||
DB018070.1 | - | 5, 6 | 556 |
