siRNA Details
Gene Symbol: ZNF609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015042.1 | NP_055857.1 | 1 | 592 | ||
XM_011521386.2 | XP_011519688.1 | 2 | 818 | |||
XM_017022021.1 | XP_016877510.1 | 2 | 800 | |||
GenBank mRNA | BC084543.1 | - | 1 | 83 | ||
BC142647.1 | - | 2 | 816 |
