siRNA Details
Gene Symbol: ZNF609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015042.1 | NP_055857.1 | 1 | 714 | ||
XM_011521386.2 | XP_011519688.1 | 2 | 940 | |||
XM_017022021.1 | XP_016877510.1 | 2 | 922 | |||
GenBank mRNA | BC084543.1 | - | 1 | 205 | ||
BC142647.1 | - | 2 | 938 |
