siRNA Details
Gene Symbol: U2SURP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080415.1 | NP_001073884.1 | 15, 16 | 1549 | ||
NM_001320219.1 | NP_001307148.1 | 15, 16 | 1546 | |||
NM_001320220.1 | NP_001307149.1 | 14, 15 | 1505 | |||
NM_001320222.1 | NP_001307151.1 | 16, 17 | 1614 | |||
XM_005247248.4 | XP_005247305.1 | 15, 16 | 1555 | |||
XM_017006037.1 | XP_016861526.1 | 11, 12 | 1157 | |||
XM_017006038.1 | XP_016861527.1 | 14, 15 | 1441 | |||
XM_017006039.1 | XP_016861528.1 | 8, 9 | 884 | |||
GenBank mRNA | AB002330.1 | - | 15, 16 | 1448 | ||
AK057679.1 | - | 2, 3 | 1543 | |||
AK074327.1 | - | 8, 9 | 813 | |||
BC006474.1 | - | 15, 16 | 1445 | |||
BC111692.1 | - | 14, 15 | 1876 | 40028600 |
