siRNA Details
Gene Symbol: KIAA0895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100425.1 | NP_001093895.1 | 4 | 1020 | ||
NM_001199706.1 | NP_001186635.1 | 4 | 1253 | |||
NM_001199707.1 | NP_001186636.1 | 3 | 1261 | |||
NM_001199708.1 | NP_001186637.1 | 4 | 1094 | |||
NM_001300956.1 | NP_001287885.1 | 3 | 405 | |||
NM_015314.2 | NP_056129.2 | 3 | 1117 | |||
XM_005249689.3 | XP_005249746.1 | 4 | 1470 | |||
XM_005249690.4 | XP_005249747.1 | 3 | 567 | |||
GenBank mRNA | AB020702.1 | - | 3 | 1031 | ||
AK302559.1 | - | 4 | 1221 | |||
AK302632.1 | - | 2 | 197 | |||
BC028678.2 | - | 4 | 979 | 4836984 | ||
BC056191.1 | - | 3 | 1001 | 5298453 | ||
BC136362.1 | - | 3 | 1011 | 9020349 | ||
BC144032.1 | - | 4 | 1158 | 9052552 | ||
BU507941.1 | - | 3 | 307 | |||
DB060820.1 | - | 2 | 428 |
