siRNA Details
Gene Symbol: KIAA0895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100425.1 | NP_001093895.1 | 4, 5 | 1050 | ||
NM_001199706.1 | NP_001186635.1 | 4, 5 | 1283 | |||
NM_001199707.1 | NP_001186636.1 | 3, 4 | 1291 | |||
NM_001199708.1 | NP_001186637.1 | 4, 5 | 1124 | |||
NM_001300956.1 | NP_001287885.1 | 3, 4 | 435 | |||
NM_015314.2 | NP_056129.2 | 3, 4 | 1147 | |||
XM_005249689.3 | XP_005249746.1 | 4, 5 | 1500 | |||
XM_005249690.4 | XP_005249747.1 | 3, 4 | 597 | |||
GenBank mRNA | AB020702.1 | - | 3, 4 | 1061 | ||
AK293912.1 | - | 4, 5 | 1056 | |||
AK302559.1 | - | 4, 5 | 1251 | |||
AK302632.1 | - | 2, 3 | 227 | |||
BC028678.2 | - | 4, 5 | 1009 | 4836984 | ||
BC056191.1 | - | 3, 4 | 1031 | 5298453 | ||
BC136362.1 | - | 3, 4 | 1041 | 9020349 | ||
BC144032.1 | - | 4, 5 | 1188 | 9052552 | ||
BU507941.1 | - | 3, 4 | 337 | |||
DB060820.1 | - | 2, 3 | 458 |
