siRNA Details
Gene Symbol: TMEM131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015348.1 | NP_056163.1 | 22, 23 | 2633 | ||
XM_005263909.1 | XP_005263966.1 | 23, 24 | 2719 | |||
XM_005263910.1 | XP_005263967.1 | 23, 24 | 2719 | |||
XM_005263911.1 | XP_005263968.1 | 22, 23 | 2602 | |||
XM_005263912.3 | XP_005263969.1 | 21, 22 | 2807 | |||
XM_011510869.1 | XP_011509171.1 | 23, 24 | 2719 | |||
GenBank mRNA | D87446.1 | - | 21, 22 | 2171 |
