siRNA Details
Gene Symbol: PLXNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012401.3 | NP_036533.2 | 32 | 5193 | ||
XM_005261909.1 | XP_005261966.1 | 33 | 5241 | |||
XM_005261910.2 | XP_005261967.1 | 33 | 5252 | |||
XM_005261911.4 | XP_005261968.1 | 32 | 5191 | |||
XM_006724413.2 | XP_006724476.1 | 34 | 5311 | |||
XM_011530682.2 | XP_011528984.1 | 33 | 5392 | |||
XM_017028703.1 | XP_016884192.1 | 34 | 5292 | |||
XM_017028704.1 | XP_016884193.1 | 33 | 5245 | |||
GenBank mRNA | AB002313.1 | - | 31 | 5110 | ||
AB075511.1 | - | 17 | 2303 | |||
AK025415.1 | - | 7 | 780 | |||
AK025701.1 | - | 19 | 2561 | |||
AK056543.1 | - | 15 | 2362 | |||
AK074932.1 | - | 14 | 1829 | |||
AK123131.1 | - | 4 | 774 | |||
AK126394.1 | - | 13 | 1902 | |||
BC004542.2 | - | 9 | 1012 | |||
BC110366.1 | - | 4 | 430 | |||
BT006887.1 | - | 9 | 996 |
