siRNA Details
Gene Symbol: TMEM158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015444.2 | NP_056259.2 | 1 | 1429 | ||
GenBank mRNA | AF438313.1 | - | 1 | 1369 | ||
AL080235.1 | - | 1 | 684 | |||
BC057390.1 | - | 2 | 1343 | 6175193 | ||
BQ000094.1 | - | 1 | 351 | |||
BU627971.1 | - | 1 | 445 |
