siRNA Details
Gene Symbol: POM121L8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024583.1 | 7 | 4830 | |||
GenBank mRNA | AL117485.1 | - | 5 | 2781 | ||
BC050331.1 | - | 2 | 1014 | |||
BC068032.1 | - | 2 | 875 | |||
BC090873.1 | - | 2 | 735 |
Gene Symbol: LOC102725072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135922.1 | 6 | 3769 | |||
GenBank mRNA | BC051721.1 | - | 6 | 3769 |
Gene Symbol: POM121L10P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024593.1 | 6 | 7968 | |||
GenBank mRNA | BX648116.1 | - | 5 | 3060 |
Gene Symbol: POM121L1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024591.1 | 10 | 3488 |
Gene Symbol: POM121L9P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003714.1 | 7 | 3720 | |||
GenBank mRNA | AL045566.1 | - | 1 | 77 | ||
AL117401.1 | - | 7 | 3514 | |||
AL117484.1 | - | 4 | 1916 | |||
BC063879.1 | - | 2 | 1217 | |||
BC086868.1 | - | 3 | 2042 |
