siRNA Details
Gene Symbol: POM121L8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024583.1 | 7 | 5490 | |||
GenBank mRNA | AL117485.1 | - | 5 | 3442 | ||
BC050331.1 | - | 2 | 1675 | |||
BC068032.1 | - | 2 | 1536 | |||
BC090873.1 | - | 2 | 1396 |
Gene Symbol: LOC102725072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135922.1 | 6 | 4430 | |||
GenBank mRNA | BC051721.1 | - | 6 | 4430 |
