siRNA Details
Gene Symbol: LOC102725072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135922.1 | 6 | 4819 | |||
GenBank mRNA | BC051721.1 | - | 6 | 4819 |
Gene Symbol: MAPKAPK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020134.1 | XP_016875623.1 | 1 | 25 |
Gene Symbol: POM121L8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024583.1 | 7 | 5878 | |||
GenBank mRNA | BC034793.1 | - | 1 | 38 | ||
BC050331.1 | - | 2 | 2065 | |||
BC050342.1 | - | 1 | 306 | |||
BC068032.1 | - | 2 | 1925 | |||
BC090873.1 | - | 2 | 1785 |
