siRNA Details
Gene Symbol: KCNK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317090.1 | NP_001304019.1 | 4 | 576 | ||
NM_033310.2 | NP_201567.1 | 4 | 713 | |||
NR_133660.1 | 3 | 466 | ||||
NR_133661.1 | 4 | 521 | ||||
GenBank mRNA | AF248242.1 | - | 3 | 750 | ||
AF259500.1 | - | 4 | 631 | |||
AK296021.1 | - | 2 | 465 | |||
AK309443.1 | - | 2 | 583 | |||
BC033577.1 | - | 4 | 521 | |||
BC110327.1 | - | 4 | 576 | |||
EU978935.1 | - | 3 | 409 |
Gene Symbol: KCNK4-TEX40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_133662.1 | 4 | 531 |
