siRNA Details
Gene Symbol: ZNF117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015852.3 | NP_056936.2 | 4 | 1391 | ||
GenBank mRNA | BC034021.1 | - | 5 | 544 | ||
M27879.1 | - | 3 | 212 | |||
M55422.1 | - | 4 | 1390 |
