siRNA Details
Gene Symbol: TMEM138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016464.4 | NP_057548.1 | 3 | 793 | ||
NR_028473.1 | 4 | 676 | ||||
XM_006718585.3 | XP_006718648.1 | 3 | 433 | |||
XM_006718586.2 | XP_006718649.1 | 3 | 433 | |||
XM_006718588.3 | XP_006718651.1 | 3 | 288 | |||
XM_011545098.2 | XP_011543400.1 | 3 | 433 | |||
XM_011545099.1 | XP_011543401.1 | 3 | 433 | |||
XM_011545100.1 | XP_011543402.1 | 3 | 433 | |||
XM_017017917.1 | XP_016873406.1 | 3 | 433 | |||
GenBank mRNA | AF151030.1 | - | 3 | 410 | ||
AF151032.1 | - | 3 | 407 | |||
AK027724.1 | - | 4 | 396 | |||
AK303219.1 | - | 3 | 408 | |||
AK307371.1 | - | 4 | 307 | |||
AL832879.1 | - | 3 | 643 | |||
BC005201.1 | - | 3 | 408 | 3684390 | ||
CB956757.1 | - | 4 | 348 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: BC132574, BC145197, AK050471, BC107026, BC058237, and others.
Gene Ontology Categories:
