siRNA Details
Gene Symbol: ZNF44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164276.1 | NP_001157748.1 | 5 | 607 | ||
NM_016264.3 | NP_057348.3 | 4 | 463 | |||
XM_005259934.3 | XP_005259991.1 | 4 | 473 | |||
XM_011528062.2 | XP_011526364.1 | 5 | 572 | |||
XM_011528064.1 | XP_011526366.1 | 4 | 888 | |||
XM_011528066.1 | XP_011526368.1 | 3 | 761 | |||
XM_017026870.1 | XP_016882359.1 | 4 | 567 | |||
XM_017026871.1 | XP_016882360.1 | 5 | 606 | |||
GenBank mRNA | AB021642.1 | - | 4 | 398 | ||
AK297528.1 | - | 5 | 521 | |||
BC136502.1 | - | 5 | 502 | 9020491 | ||
CN364680.1 | - | 4 | 442 | |||
DA815414.1 | - | 4 | 366 |
