siRNA Details
Gene Symbol: ZNF586 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204814.1 | NP_001191743.1 | 3 | 325 | ||
NM_017652.3 | NP_060122.2 | 2 | 275 | |||
GenBank mRNA | AB074270.1 | - | 2 | 258 | ||
AK000077.1 | - | 2 | 257 | |||
AK095993.1 | - | 3 | 323 | |||
AK302151.1 | - | 3 | 231 | |||
BC027922.1 | - | 2 | 261 | |||
DA904413.1 | - | 2 | 270 |
