siRNA Details
Gene Symbol: ZNF586 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077426.2 | NP_001070894.1 | 2 | 281 | ||
NM_001204814.1 | NP_001191743.1 | 4 | 458 | |||
NM_017652.3 | NP_060122.2 | 3 | 408 | |||
GenBank mRNA | AB074270.1 | - | 3 | 391 | ||
AK000077.1 | - | 3 | 390 | |||
AK095993.1 | - | 4 | 456 | |||
AK300419.1 | - | 2 | 238 | |||
AK302151.1 | - | 4 | 364 | |||
BC124558.1 | - | 2 | 219 | 40085152 | ||
BP352135.1 | - | 2 | 282 | |||
DA904413.1 | - | 3 | 403 |
