siRNA Details
Gene Symbol: LOC55338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130914.1 | 3 | 650 | |||
GenBank mRNA | AK002104.1 | - | 2 | 1344 | ||
AK002154.1 | - | 2 | 571 | |||
AK024354.1 | - | 1 | 297 | |||
BC117434.1 | - | 2 | 456 |
