siRNA Details
Gene Symbol: AJAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042478.1 | NP_001035943.1 | 1, 2 | 413 | ||
NM_018836.3 | NP_061324.1 | 1, 2 | 413 | |||
XM_011541786.2 | XP_011540088.1 | 1, 2 | 683 | |||
GenBank mRNA | AF175409.1 | - | 1, 2 | 725 | ||
AY282806.1 | - | 1, 2 | 413 |
