siRNA Details
Gene Symbol: KIAA1211 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020722.1 | NP_065773.1 | 7 | 868 | ||
XM_005265752.2 | XP_005265809.1 | 6 | 875 | |||
XM_005265755.2 | XP_005265812.1 | 3 | 304 | |||
XM_005265756.2 | XP_005265813.1 | 3 | 352 | |||
XM_006714045.2 | XP_006714108.1 | 3 | 329 | |||
XM_011534397.2 | XP_011532699.1 | 6 | 1618 | |||
XM_017008473.1 | XP_016863962.1 | 6 | 604 | |||
XM_017008474.1 | XP_016863963.1 | 5 | 1482 | |||
GenBank mRNA | AK097530.1 | - | 3 | 329 | ||
AK126014.1 | - | 7 | 868 | |||
AK302486.1 | - | 3 | 352 | |||
BC144536.1 | - | 6 | 604 | 9053063 | ||
BC150615.1 | - | 6 | 604 | 9056985 | ||
BC171783.1 | - | 6 | 604 | 9054437 |
