siRNA Details
Gene Symbol: CARD18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021571.3 | NP_067546.1 | 1 | 76 | ||
GenBank mRNA | AF208005.1 | - | 1 | 68 | ||
AI189838.1 | - | 2 | 360 | |||
AY358231.1 | - | 1 | 80 | |||
BC131610.1 | - | 1 | 68 | 40115072 |
