siRNA Details
Gene Symbol: ANAPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022662.3 | NP_073153.1 | 19 | 2402 | ||
XM_011511633.2 | XP_011509935.1 | 19 | 2434 | |||
XM_011511634.2 | XP_011509936.1 | 19 | 2549 | |||
XM_011511636.2 | XP_011509938.1 | 19 | 2434 | |||
XM_011511637.2 | XP_011509939.1 | 19 | 2434 | |||
XM_017004710.1 | XP_016860199.1 | 19 | 2434 | |||
XM_017004711.1 | XP_016860200.1 | 19 | 2434 | |||
XM_017004712.1 | XP_016860201.1 | 19 | 2549 | |||
XM_017004713.1 | XP_016860202.1 | 18 | 2497 | |||
XM_017004714.1 | XP_016860203.1 | 18 | 2445 | |||
XM_017004715.1 | XP_016860204.1 | 19 | 2434 | |||
GenBank mRNA | AJ278357.1 | - | 19 | 2417 | ||
AK023807.1 | - | 6 | 628 | |||
AL832013.1 | - | 8 | 758 | |||
BC104902.1 | - | 19 | 2204 | 8143905 | ||
BC104904.1 | - | 19 | 2204 | 8143907 |
