siRNA Details
Gene Symbol: PLPPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022737.2 | NP_073574.2 | 3, 4 | 433 | ||
XM_005260017.1 | XP_005260074.1 | 3, 4 | 433 | |||
XM_017027147.1 | XP_016882636.1 | 3, 4 | 436 | |||
GenBank mRNA | AK023117.1 | - | 3, 4 | 394 | ||
AK225482.1 | - | 3, 4 | 348 | |||
AY304516.1 | - | 3, 4 | 388 | |||
AY339628.1 | - | 2, 3 | 88 | |||
BC009378.1 | - | 3, 4 | 276 | 4125316 |
