siRNA Details
Gene Symbol: STAG3L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040585.1 | 4 | 595 | |||
NR_040586.1 | 3 | 519 | ||||
GenBank mRNA | AK023257.1 | - | 3 | 513 | ||
AK307290.1 | - | 4 | 575 | |||
AK308931.1 | - | 4 | 377 | |||
BC026058.2 | - | 3 | 485 | 3936696 | ||
CR457306.1 | - | 2 | 261 |
