siRNA Details
Gene Symbol: PVRIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024070.3 | NP_076975.2 | 3 | 635 | ||
XM_011516575.2 | XP_011514877.1 | 4 | 651 | |||
GenBank mRNA | BC001129.2 | - | 2 | 327 | ||
BC032113.2 | - | 2 | 283 | |||
BC073861.1 | - | 2 | 298 | |||
BC107873.1 | - | 4 | 660 | 6452536 | ||
BC112090.1 | - | 2 | 363 | |||
BC112092.1 | - | 2 | 364 |
