siRNA Details
Gene Symbol: C1orf35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024319.3 | NP_077295.1 | 1 | 299 | ||
NR_130975.1 | 1 | 299 | ||||
GenBank mRNA | AK123377.1 | - | 1 | 279 | ||
AK126087.1 | - | 1 | 2248 | |||
AY137773.1 | - | 1 | 160 | |||
BC002843.2 | - | 1 | 158 | 3634226 | ||
BC062585.1 | - | 1 | 155 | 5212995 |
