siRNA Details
Gene Symbol: LINC00472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026807.2 | 1 | 500 | |||
NR_121612.1 | 1 | 500 | ||||
NR_121613.1 | 1 | 500 | ||||
NR_121614.1 | 1 | 500 | ||||
GenBank mRNA | AI498652.1 | - | 1 | 22 | ||
AK023251.1 | - | 1 | 500 | |||
CD674415.1 | - | 1 | 16 |
