siRNA Details
Gene Symbol: LINC00472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026807.2 | 1 | 585 | |||
NR_121612.1 | 1 | 585 | ||||
NR_121613.1 | 1 | 585 | ||||
NR_121614.1 | 1 | 585 | ||||
GenBank mRNA | AI498652.1 | - | 1 | 107 | ||
AI985403.1 | - | 1 | 107 | |||
AK023251.1 | - | 1 | 585 | |||
AW274058.1 | - | 1 | 91 | |||
CD674415.1 | - | 1 | 101 |
