siRNA Details
Gene Symbol: FAM106A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026809.1 | 1 | 1431 | |||
GenBank mRNA | AK021862.1 | - | 1 | 1431 | ||
BC098263.1 | - | 1 | 1410 |
Gene Symbol: FAM106CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC098294.1 | - | 1 | 1410 |
