siRNA Details
Gene Symbol: SMG1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135317.1 | 1 | 254 | |||
GenBank mRNA | AB055226.1 | - | 1 | 256 |
Gene Symbol: SLC7A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB017908.1 | - | 1 | 217 |
