siRNA Details
Gene Symbol: ZNF469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127464.2 | NP_001120936.2 | 2 | 6015 | ||
XM_017023784.1 | XP_016879273.1 | 2 | 7249 | |||
XM_017023785.1 | XP_016879274.1 | 2 | 6292 | |||
GenBank mRNA | AB058761.2 | - | 1 | 1649 |
