siRNA Details
Gene Symbol: KIAA1671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145206.1 | NP_001138678.1 | 8 | 5139 | ||
XM_005261793.2 | XP_005261850.1 | 10 | 5452 | |||
XM_005261794.4 | XP_005261851.1 | 10 | 5405 | |||
XM_006724346.2 | XP_006724409.1 | 11 | 5391 | |||
GenBank mRNA | AB051458.1 | - | 5 | 787 | ||
AL832019.1 | - | 5 | 4521 | |||
BC171801.1 | - | 8 | 5139 | 9054455 |
