siRNA Details
Gene Symbol: ZNF518B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053042.2 | NP_444270.2 | 3 | 2231 | ||
XM_005248193.2 | XP_005248250.1 | 2 | 2066 | |||
XM_017008784.1 | XP_016864273.1 | 2 | 4689 | |||
XM_017008785.1 | XP_016864274.1 | 3 | 2457 | |||
XM_017008786.1 | XP_016864275.1 | 2 | 2291 | |||
GenBank mRNA | AB051516.1 | - | 1 | 1583 | ||
AK058072.2 | - | 1 | 422 | |||
AK122755.1 | - | 1 | 918 | |||
AL833306.1 | - | 1 | 1767 |
