siRNA Details
Gene Symbol: OR6W1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002140.1 | 1 | 866 | |||
GenBank mRNA | BC069382.1 | - | 1 | 457 | ||
BC074961.2 | - | 1 | 442 | |||
BC074962.2 | - | 1 | 442 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR6W1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002140.1 | 1 | 866 | |||
GenBank mRNA | BC069382.1 | - | 1 | 457 | ||
BC074961.2 | - | 1 | 442 | |||
BC074962.2 | - | 1 | 442 |