siRNA Details
Gene Symbol: ST7-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002331.3 | 6 | 629 | |||
NR_109981.1 | 5 | 525 | ||||
GenBank mRNA | AF400045.1 | - | LIT1899 ST7OT2 |
6 | 619 | |
DB054726.1 | - | 5 | 525 |
