siRNA Details
Gene Symbol: LRRC4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080457.1 | NP_001073926.1 | 3 | 1276 | ||
XM_005259429.4 | XP_005259486.1 | 3 | 1455 | |||
XM_006723506.3 | XP_006723569.1 | 4 | 1557 | |||
XM_011527520.2 | XP_011525822.1 | 3 | 2703 | |||
GenBank mRNA | AL137451.1 | - | 1 | 496 | ||
BC019687.3 | - | 1 | 520 | |||
BC032460.1 | - | 2 | 1151 | |||
BC131897.1 | - | 3 | 1254 |
