siRNA Details
Gene Symbol: MRFAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203462.2 | NP_982287.1 | 1 | 365 | ||
XM_005247932.1 | XP_005247989.1 | 1 | 351 | |||
GenBank mRNA | AF155654.1 | - | 1 | 363 | ||
AF258591.1 | - | 1 | 313 | |||
AK312676.1 | - | 1 | 344 | |||
BC008087.1 | - | 1 | 312 | 3446352 | ||
KC858143.1 | - | 1 | 118 |
