siRNA Details
Gene Symbol: MRFAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203462.2 | NP_982287.1 | 1 | 525 | ||
XM_005247932.1 | XP_005247989.1 | 1 | 511 | |||
GenBank mRNA | AF258591.1 | - | 1 | 473 | ||
AK312676.1 | - | 1 | 504 | |||
BC008087.1 | - | 1 | 472 | 3446352 | ||
BC066897.1 | - | 1 | 525 | 4818881 | ||
KC858143.1 | - | 1 | 278 |
