siRNA Details
Gene Symbol: KCTD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138444.3 | NP_612453.1 | 1 | 1193 | ||
GenBank mRNA | AF359381.1 | - | 1 | 1178 | ||
AK128363.1 | - | 1 | 1196 | |||
BC013764.1 | - | 1 | 1023 | 4521096 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_177715, AK035535, BC116648, AK173260, AK172581, and others.
Gene Ontology Categories:
