siRNA Details
Gene Symbol: FAM186A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145475.1 | NP_001138947.1 | 4 | 1560 | ||
XM_006719231.3 | XP_006719294.1 | 4 | 1571 | |||
XM_011537890.2 | XP_011536192.1 | 4 | 1571 | |||
XM_011537892.1 | XP_011536194.1 | 3 | 1080 | |||
GenBank mRNA | AL833333.1 | - | 4 | 1560 |
