siRNA Details
Gene Symbol: C19orf70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308240.1 | NP_001295169.1 | 4 | 835 | ||
NM_205767.2 | NP_991330.1 | 3 | 639 | |||
XM_011527675.2 | XP_011525977.1 | 3 | 588 | |||
XM_017026248.1 | XP_016881737.1 | 3 | 580 | |||
GenBank mRNA | AA292888.1 | - | 3 | 406 | ||
AI300708.1 | - | 2 | 264 | |||
AJ707830.1 | - | 3 | 385 | |||
AK091768.1 | - | 1 | 1527 | |||
BC009557.1 | - | 3 | 273 | 3899541 | ||
BC042386.1 | - | 3 | 248 | 5288954 | ||
BC052640.1 | - | 3 | 250 | |||
BI827162.1 | - | 3 | 638 |
