siRNA Details
Gene Symbol: LOC101929715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110597.1 | 2 | 354 | |||
GenBank mRNA | AW303360.1 | - | 3 | 171 | ||
BC111372.1 | - | 1 | 601 | |||
HY015289.1 | - | 1 | 23 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110597.1 | 2 | 354 | |||
GenBank mRNA | AW303360.1 | - | 3 | 171 | ||
BC111372.1 | - | 1 | 601 | |||
HY015289.1 | - | 1 | 23 |