siRNA Details
Gene Symbol: C7orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100159.2 | NP_001093629.1 | 7 | 872 | ||
XM_006715649.3 | XP_006715712.1 | 6 | 732 | |||
XM_006715650.2 | XP_006715713.1 | 6 | 699 | |||
XM_011515121.1 | XP_011513423.1 | 7 | 776 | |||
GenBank mRNA | BC031107.1 | - | 7 | 863 |
