siRNA Details
Gene Symbol: TLDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304783.1 | NP_001291712.1 | 2, 3 | 237 | ||
NM_080628.2 | NP_542195.1 | 2, 3 | 237 | |||
XM_017027672.1 | XP_016883161.1 | 2, 3 | 227 | |||
XM_017027673.1 | XP_016883162.1 | 3, 4 | 352 | |||
GenBank mRNA | AK123544.1 | - | 2, 3 | 191 | ||
BC127688.1 | - | 2, 3 | 237 | 40127440 | ||
BC130646.1 | - | 2, 3 | 226 | 40146677 | ||
BC130648.1 | - | 2, 3 | 226 | 40146679 | ||
BY994247.1 | - | 2, 3 | 225 |
