siRNA Details
Gene Symbol: LINC01620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132342.1 | 2 | 269 | |||
GenBank mRNA | BC030259.1 | - | 2 | 270 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132342.1 | 2 | 269 | |||
GenBank mRNA | BC030259.1 | - | 2 | 270 |