siRNA Details
Gene Symbol: VTI1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006717637.2 | XP_006717700.1 | 9 | 2132 | ||
XM_011539331.2 | XP_011537633.1 | 9 | 2143 | |||
XM_017015748.1 | XP_016871237.1 | 8 | 2065 |
Gene Symbol: LOC143188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK026759.1 | - | 1 | 1023 | ||
AK074471.1 | - | 2 | 2578 | |||
BC093709.1 | - | 1 | 173 | |||
BC112004.1 | - | 1 | 173 |
