siRNA Details
Gene Symbol: C15orf61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143936.1 | NP_001137408.1 | 1 | 254 | ||
GenBank mRNA | AK294012.1 | - | 1 | 189 | ||
AY726587.1 | - | 1 | 146 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C15orf61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143936.1 | NP_001137408.1 | 1 | 254 | ||
GenBank mRNA | AK294012.1 | - | 1 | 189 | ||
AY726587.1 | - | 1 | 146 |